Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-Benzyl pyrrolidin-3-ylcarbamate hydrochloride 97%

Supplier: AMBEED, INC
Warning

A105115-1G A105115-250MG A105115-5G
76970-378EA 7.43 USD InStock
76970-378 76970-380 76970-382 76970-384
(S)-Benzyl pyrrolidin-3-ylcarbamate hydrochloride 97%
(S)-Benzyl pyrrolidin-3-ylcarbamate hydrochloride
Precaución: for Research Use Only


Formula: C₁₂H₁₇ClN₂O₂
MDL Number: MFCD09749851
CAS Number: 1217631-74-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty