Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Benzyltriethylammonium chloride 97%

Supplier: AMBEED, INC
Warning

A114975-1000G A114975-100G A114975-25G A114975-500G
76971-112EA 15 USD InStock
76971-112 76971-114 76971-116 76971-118
Benzyltriethylammonium chloride 97%
Benzyltriethylammonium chloride
Precaución: for Research Use Only


Formula: C₁₃H₂₂ClN
MW: 227.78 g/mol
MDL Number: MFCD00011824
CAS Number: 56-37-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty