Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Cyclopropylpiperidin-3-amine 95%

Supplier: AMBEED, INC
Warning

A287432-100MG A287432-1G A287432-250MG
76977-518EA 44 USD InStock
76977-518 76977-520 76977-522
N-Cyclopropylpiperidin-3-amine 95%
N-Cyclopropylpiperidin-3-amine
Precaución: for Research Use Only


Formula: C₈H₁₆N₂
MDL Number: MFCD16066355
CAS Number: 1251083-96-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty