Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Hydroxy-3-nitro-5-(trifluoromethyl)pyridine 98%

Supplier: AMBEED, INC
Warning

A107577-100G A107577-10G A107577-1G A107577-250MG A107577-25G A107577-5G
76970-518EA 624 USD InStock
76970-518 76970-520 76970-522 76970-524 76970-526 76970-528
2-Hydroxy-3-nitro-5-(trifluoromethyl)pyridine 98%
2-Hydroxy-3-nitro-5-(trifluoromethyl)pyridine
Precaución: for Research Use Only


Formula: C₆H₃F₃N₂O₃
MDL Number: MFCD00153193
CAS Number: 33252-64-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty