Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ranolazine dihydrochloride 98%

Supplier: AMBEED, INC
Warning

A730331-100MG A730331-1G A730331-250MG A730331-5G
76981-270EA 19 USD InStock
76981-270 76981-272 76981-274 76981-276
Ranolazine dihydrochloride 98%
Ranolazine dihydrochloride
Precaución: for Research Use Only


Formula: C₂₄H₃₃N₃O₄
MDL Number: MFCD03788770
CAS Number: 95635-56-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty