Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Chloro-6-fluoro-3-nitrotoluene 95%

Supplier: AMBEED, INC
Warning

A445490-1G A445490-250MG A445490-5G
76979-426EA 10 USD InStock
76979-426 76979-428 76979-430
2-Chloro-6-fluoro-3-nitrotoluene 95%
2-Chloro-6-fluoro-3-nitrotoluene
Precaución: for Research Use Only


Formula: C₇H₅ClFNO₂
MDL Number: MFCD21603723
CAS Number: 90292-62-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty