Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Pyridine-3,4,5-triamine 97%

Supplier: AMBEED, INC
Warning

A224037-100MG A224037-1G A224037-250MG
76975-916EA 33 USD InStock
76975-916 76975-918 76975-920
Pyridine-3,4,5-triamine 97%
Pyridine-3,4,5-triamine
Precaución: for Research Use Only


Formula: C₅H₈N₄
MDL Number: MFCD09753793
CAS Number: 618439-82-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty