Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Uridine 5′-monophosphate disodium salt 98%

Supplier: AMBEED, INC
Warning

A260727-25G A260727-25MG A260727-5G A260727-100G A260727-500G A260727-1000G
76976-942EA 16 USD InStock
76976-942 76976-944 76976-946 77601-098 77672-328 77672-326
Uridine 5′-monophosphate disodium salt 98%
Uridine 5′-monophosphate disodium salt
Precaución: for Research Use Only


Formula: C₉H₁₁N₂Na₂O₉P
MW: 368.15 g/mol
MDL Number: MFCD00006525
CAS Number: 3387-36-8
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty