Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Cyclopentanecarboxylic acid 98%

Supplier: AMBEED, INC
Warning

A312068-100G A312068-25G A312068-500G A312068-5G
76977-930EA 65 USD InStock
76977-930 76977-932 76977-934 76977-936
Cyclopentanecarboxylic acid 98%
Cyclopentanecarboxylic acid
Precaución: for Research Use Only


Formula: C₆H₁₀O₂
MDL Number: MFCD00001371
CAS Number: 3400-45-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty