Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Boc-D-glutamic acid 1-benzyl ester 98%

Supplier: AMBEED, INC
Warning

A111983-1G A111983-10G A111983-5G
76970-886EA 6 USD InStock
76970-886 77670-582 77670-584
N-Boc-D-glutamic acid 1-benzyl ester 98%
N-Boc-D-glutamic acid 1-benzyl ester
Precaución: for Research Use Only


Formula: C₁₇H₂₃NO₆
MW: 337.37 g/mol
MDL Number: MFCD00038266
CAS Number: 34404-30-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty