Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate 98%

Supplier: AMBEED, INC
Warning

A616943-100G A616943-1G A616943-25G A616943-500G A616943-5G
76980-588EA 109 USD InStock
76980-588 76980-590 76980-592 76980-594 76980-596
1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate 98%
1,1'-Binaphthyl-2,2'-diyl hydrogen phosphate
Precaución: for Research Use Only


Formula: C₂₀H₁₃O₄P
MDL Number: MFCD00010045
CAS Number: 35193-63-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty