Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Ethyl-4-(4-piperidinyloxy)benzoate hydrochloride 95%

Supplier: AMBEED, INC
Warning

A560231-100MG A560231-1G A560231-250MG
76980-266EA 31 USD InStock
76980-266 76980-268 76980-270
Ethyl-4-(4-piperidinyloxy)benzoate hydrochloride 95%
Ethyl-4-(4-piperidinyloxy)benzoate hydrochloride
Precaución: for Research Use Only


Formula: C₁₄H₂₀ClNO₃
MW: 285.77 g/mol
MDL Number: MFCD13560942
CAS Number: 950649-15-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty