Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

N-Acetoacetylanthranilic acid hydrate 95+%

Supplier: AMBEED, INC
Warning

A502623-1000G A502623-100G A502623-500G
76979-884EA 108 USD InStock
76979-884 76979-886 76979-888
N-Acetoacetylanthranilic acid hydrate 95+%
N-Acetoacetylanthranilic acid hydrate
Precaución: for Research Use Only


Formula: C₁₁H₁₁NO₄
MDL Number: MFCD00089988
CAS Number: 35354-86-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty