Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,5-Dimethylpiperidin (mixture of cis- and trans- isomers) 98%

Supplier: AMBEED, INC
Danger

A878859-10G A878859-25G
76981-866EA 11 USD InStock
76981-866 76981-868 76981-870
3,5-Dimethylpiperidin (mixture of cis- and trans- isomers) 98%
3,5-Dimethylpiperidin (mixture of cis- and trans-isomers)
Precaución: for Research Use Only


Formula: C₇H₁₅N
MDL Number: MFCD00005996
CAS Number: 35794-11-7
UN: 1993
ADR: 3,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty