Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Chloro-4'-methoxypropiophenone 95%

Supplier: AMBEED, INC
Warning

A537633-100MG A537633-1G A537633-250MG A537633-5G
76980-134EA 15 USD InStock
76980-134 76980-136 76980-138 76980-140
3-Chloro-4'-methoxypropiophenone 95%
3-Chloro-4'-methoxypropiophenone
Precaución: For research use only.


Formula: C₁₀H₁₁ClO₂
MDL Number: MFCD00236602
CAS Number: 35999-20-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty