Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-tert-Butyl-1,4-benzoquinone 98%

Supplier: AMBEED, INC
Warning

A254740-100G A254740-1G A254740-25G A254740-5G
76976-760EA 108 USD InStock
76976-760 76976-762 76976-764 76976-766
2-tert-Butyl-1,4-benzoquinone 98%
2-tert-Butyl-1,4-benzoquinone
Precaución: for Research Use Only


Formula: C₁₀H₁₂O₂
MDL Number: MFCD00666928
CAS Number: 3602-55-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty