Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2,5-Dichloro-4-(trifluoromethyl)pyrimidine 95%

Supplier: AMBEED, INC
Warning

A399081-100MG A399081-1G A399081-250MG A399081-5G
76979-040EA 19 USD InStock
76979-040 76979-042 76979-044 76979-046
2,5-Dichloro-4-(trifluoromethyl)pyrimidine 95%
2,5-Dichloro-4-(trifluoromethyl)pyrimidine
Precaución: for Research Use Only


Formula: C₅HCl₂F₃N₂
MW: 216.98 g/mol
MDL Number: MFCD13176599
CAS Number: 785777-98-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty