Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Amino-4H-1,2,4-triazole-3-carboxylic acid 95%

Supplier: AMBEED, INC
Warning

A112972-100G A112972-10G A112972-1G A112972-25G A112972-5G
76970-958EA 58 USD InStock
76970-958 76970-960 76970-962 76970-964 76970-966
5-Amino-4H-1,2,4-triazole-3-carboxylic acid 95%
5-Amino-4H-1,2,4-triazole-3-carboxylic acid
Precaución: for Research Use Only


Formula: C₃H₄N₄O₂
MDL Number: MFCD00010620
CAS Number: 3641-13-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty