Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-[Ethyl-(4-piperidinyl)amino]-1-ethanol dihydrochloride 95%

Supplier: AMBEED, INC
Warning

A591119-100MG A591119-1G A591119-250MG
76980-440EA 12 USD InStock
76980-440 76980-442 76980-444
2-[Ethyl-(4-piperidinyl)amino]-1-ethanol dihydrochloride 95%
2-[Ethyl-(4-piperidinyl)amino]-1-ethanol dihydrochloride
Precaución: for Research Use Only


Formula: C₉H₂₂Cl₂N₂O
MDL Number: MFCD13561566
CAS Number: 1219979-66-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty