Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

D-Tyrosine methyl ester hydrochloride 98%

Supplier: AMBEED, INC
Warning

A218471-100G A218471-10G A218471-25G A218471-5G
76975-760EA 83 USD InStock
76975-760 76975-762 76975-764 76975-766
D-Tyrosine methyl ester hydrochloride 98%
D-Tyrosine methyl ester hydrochloride
Precaución: for Research Use Only


Formula: C₁₀H₁₄ClNO₃
MDL Number: MFCD00066124
CAS Number: 3728-20-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty