Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-6-hydroxy-2-methylpyridine 95%

Supplier: AMBEED, INC
Warning

A131401-100MG A131401-1G A131401-250MG
76972-084EA 12 USD InStock
76972-084 76972-086 76972-088
5-Bromo-6-hydroxy-2-methylpyridine 95%
5-Bromo-6-hydroxy-2-methylpyridine
Precaución: for Research Use Only


Formula: C₆H₆BrNO
MW: 188.022 g/mol
MDL Number: MFCD03095217
CAS Number: 374633-33-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty