Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4,6-Dichloro-1,3,5-triazin-2-amine 97%

Supplier: AMBEED, INC
Warning

A135461-100G A135461-1G A135461-250MG A135461-5G A135461-25G
76972-322EA 223 USD InStock
76972-322 76972-324 76972-326 76972-328 77599-968
4,6-Dichloro-1,3,5-triazin-2-amine 97%
4,6-Dichloro-1,3,5-triazin-2-amine
Precaución: For research use only.


Formula: C₃H₂Cl₂N₄
MDL Number: MFCD00078331
CAS Number: 933-20-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty