Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Fluoroisoquinoline 97%

Supplier: AMBEED, INC
Warning

A327467-1G A327467-250MG A327467-5G
76978-186EA 36 USD InStock
76978-186 76978-188 76978-190
4-Fluoroisoquinoline 97%
4-Fluoroisoquinoline
Precaución: for Research Use Only


Formula: C₉H₆FN
MDL Number: MFCD06656128
CAS Number: 394-67-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty