Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

D-Pentafluorophenylalanine 96%

Supplier: AMBEED, INC
Warning

A132698-1G A132698-250MG A132698-5G
76972-158EA 90 USD InStock
76972-158 76972-160 76972-162
D-Pentafluorophenylalanine 96%
D-Pentafluorophenylalanine
Precaución: for Research Use Only


Formula: C₉H₆F₅NO₂
MDL Number: MFCD01860872
CAS Number: 40332-58-9
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty