Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(Methoxymethyl)benzoic acid 95%

Supplier: AMBEED, INC
Warning

A301668-100MG A301668-1G A301668-250MG A301668-5G
76977-738EA 24 USD InStock
76977-738 76977-740 76977-742 76977-744
2-(Methoxymethyl)benzoic acid 95%
2-(Methoxymethyl)benzoic acid
Precaución: for Research Use Only


Formula: C₉H₁₀O₃
MW: 166.18 g/mol
MDL Number: MFCD09863839
CAS Number: 88550-19-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty