Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl-2-chloro-3-quinolinecarboxylate 98%

Supplier: AMBEED, INC
Warning

A987906-100MG A987906-1G A987906-250MG A987906-5G
76982-060EA 7 USD InStock
76982-060 76982-062 76982-064 76982-066
Methyl-2-chloro-3-quinolinecarboxylate 98%
Methyl-2-chloro-3-quinolinecarboxylate
Precaución: for Research Use Only


Formula: C₁₁H₈ClNO₂
MW: 221.64 g/mol
MDL Number: MFCD06203870
CAS Number: 16498-85-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty