Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

5-Bromo-6-hydroxynicotinic acid 98%

Supplier: AMBEED, INC
Warning

A284216-100G A284216-1G A284216-5G
76977-476EA 99 USD InStock
76977-476 76977-478 76977-480
5-Bromo-6-hydroxynicotinic acid 98%
5-Bromo-6-hydroxynicotinic acid
Precaución: for Research Use Only


Formula: C₆H₄BrNO₃
MW: 218.01 g/mol
MDL Number: MFCD08235173
CAS Number: 41668-13-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty