Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl (4-bromophenyl)acetate 98%

Supplier: AMBEED, INC
Warning

A223716-100G A223716-10G A223716-500G A223716-5G A223716-25G
76975-904EA 27 USD InStock
76975-904 76975-906 76975-908 76975-910 77303-666
Methyl (4-bromophenyl)acetate 98%
Methyl (4-bromophenyl)acetate
Precaución: for Research Use Only


Formula: C₉H₉BrO₂
MDL Number: MFCD00126844
CAS Number: 41841-16-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty