Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 2-bromo-3-methoxypropanoate 95%

Supplier: AMBEED, INC
Danger

A267693-100MG A267693-1G A267693-250MG A267693-5G
76977-142EA 15 USD InStock
76977-142 76977-144 76977-146 76977-148
Methyl 2-bromo-3-methoxypropanoate 95%
Methyl 2-bromo-3-methoxypropanoate
Precaución: for Research Use Only


Formula: C₅H₉BrO₃
MW: 197.027 g/mol
MDL Number: MFCD07784248
CAS Number: 27704-96-7
UN: 1993
ADR: 3 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty