Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1-Benzylpiperidine-3,5-dione 97%

Supplier: AMBEED, INC
Warning

A505473-100MG A505473-1G A505473-5G
76979-916EA 14 USD InStock
76979-916 76979-918 76979-920
1-Benzylpiperidine-3,5-dione 97%
1-Benzylpiperidine-3,5-dione
Precaución: for Research Use Only


Formula: C₁₂H₁₃NO₂
MDL Number: MFCD08669419
CAS Number: 50866-56-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty