Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

1,4-Dichloro-5,6,7,8-tetrahydrophthalazine 97%

Supplier: AMBEED, INC
Warning

A117671-100MG A117671-1G A117671-250MG A117671-5G
76971-314EA 38 USD InStock
76971-314 76971-316 76971-318 76971-320
1,4-Dichloro-5,6,7,8-tetrahydrophthalazine 97%
1,4-Dichloro-5,6,7,8-tetrahydrophthalazine
Precaución: for Research Use Only


Formula: C₈H₈Cl₂N₂
MDL Number: MFCD02940890
CAS Number: 67279-24-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty