Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Thieno[3,2-b]pyridine 97%

Supplier: AMBEED, INC
Warning

A109660-100MG A109660-1G A109660-250MG
76970-658EA 18 USD InStock
76970-658 76970-660 76970-662
Thieno[3,2-b]pyridine 97%
Thieno[3,2-b]pyridine
Precaución: for Research Use Only


Formula: C₇H₅NS
MDL Number: MFCD11110437
CAS Number: 272-67-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty