Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-7-bromo-1H-indazole 98%

Supplier: AMBEED, INC
Warning

A120245-10G A120245-1G A120245-250MG A120245-5G
76971-456EA 230 USD InStock
76971-456 76971-458 76971-460 76971-462
3-Amino-7-bromo-1H-indazole 98%
3-Amino-7-bromo-1H-indazole
Precaución: for Research Use Only


Formula: C₇H₆BrN₃
MDL Number: MFCD12406137
CAS Number: 1234616-28-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty