Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-1-methylpyridin-2(1H)-one 95%

Supplier: AMBEED, INC
Warning

A719557-100MG A719557-1G A719557-250MG A719557-5G
76981-230EA 18 USD InStock
76981-230 76981-232 76981-234 76981-236
3-Amino-1-methylpyridin-2(1H)-one 95%
3-Amino-1-methylpyridin-2(1H)-one
Precaución: for Research Use Only


Formula: C₆H₈N₂O
MW: 124.14 g/mol
MDL Number: MFCD12402373
CAS Number: 33631-01-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty