Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-3-methoxynaphthalene 98%

Supplier: AMBEED, INC
Warning

A159280-1G A159280-250MG A159280-25G A159280-5G
76973-464EA 17 USD InStock
76973-464 76973-466 76973-468 77567-312
2-Bromo-3-methoxynaphthalene 98%
2-Bromo-3-methoxynaphthalene
Precaución: For research use only.


Formula: C₁₁H₉BrO
MW: 237.0926 g/mol
MDL Number: MFCD11870110
CAS Number: 68251-77-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty