Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Amino-5-bromopicolinic acid 97%

Supplier: AMBEED, INC
Warning

A990985-100G A990985-10G A990985-1G A990985-250MG A990985-25G A990985-5G
76982-068EA 520 USD InStock
76982-068 76982-070 76982-072 76982-074 76982-076 76982-078
3-Amino-5-bromopicolinic acid 97%
3-Amino-5-bromopicolinic acid
Precaución: for Research Use Only


Formula: C₆H₅BrN₂O₂
MDL Number: MFCD09999770
CAS Number: 870997-85-6
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty