Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-(3-Bromophenyl)propan-2-ol 97%

Supplier: AMBEED, INC
Warning

A185319-100MG A185319-250MG A185319-25G A185319-5G A185319-1G
76974-584EA 6 USD InStock
76974-584 76974-586 76974-588 76974-590 77671-620
2-(3-Bromophenyl)propan-2-ol 97%
2-(3-Bromophenyl)propan-2-ol
Precaución: for Research Use Only


Formula: C₉H₁₁BrO
MDL Number: MFCD11870097
CAS Number: 30951-66-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty