Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(Bromomethyl)cycloheptane 98%

Supplier: AMBEED, INC
Warning

A388476-100MG A388476-250MG A388476-5G
76978-922EA 19 USD InStock
76978-922 76978-924 77672-930
(Bromomethyl)cycloheptane 98%
(Bromomethyl)cycloheptane
Precaución: for Research Use Only


Formula: C₈H₁₅Br
MDL Number: MFCD12025292
CAS Number: 3814-32-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty