Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

2-Bromo-5-fluoro-4-methylpyridine 98%

Supplier: AMBEED, INC
Warning

A138433-1G A138433-250MG A138433-10G A138433-25G
76972-458EA 14 USD InStock
76972-458 76972-460 77670-982 77670-984
2-Bromo-5-fluoro-4-methylpyridine 98%
2-Bromo-5-fluoro-4-methylpyridine
Precaución: for Research Use Only


Formula: C₆H₅BrFN
MDL Number: MFCD08277310
CAS Number: 885168-20-7
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty