Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3,3-Dimethoxycyclobutane-1-carboxylate methyl ester 98%

Supplier: AMBEED, INC
Warning

A674943-100G A674943-10G A674943-1G A674943-25G A674943-500G A674943-5G
76980-962EA 122 USD InStock
76980-962 76980-964 76980-966 76980-968 76980-970 76980-972
3,3-Dimethoxycyclobutane-1-carboxylate methyl ester 98%
3,3-Dimethoxycyclobutane-1-carboxylate methyl ester
Precaución: for Research Use Only


Formula: C₈H₁₄O₄
MDL Number: MFCD11111944
CAS Number: 98231-07-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty