Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

R406 free base 98%

Supplier: AMBEED, INC
Warning

A233735-10MG A233735-2MG A233735-5MG A233735-100MG A233735-1G A233735-250MG A233735-5G
76976-236EA 10 USD InStock
76976-236 76976-238 76976-240 77672-074 77672-076 77672-078 77672-080
R406 free base 98%
R406 free base
Precaución: for Research Use Only


Formula: C₂₂H₂₃FN₆O₅
MW: 470.45 g/mol
MDL Number: MFCD09970820
CAS Number: 841290-80-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty