Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Chloro-2-methoxypyridine-3-carboxylic acid 98%

Supplier: AMBEED, INC
Warning

A163252-100MG A163252-10G A163252-1G A163252-250MG A163252-5G
76973-658EA 22 USD InStock
76973-658 76973-660 76973-662 76973-664 76973-666
6-Chloro-2-methoxypyridine-3-carboxylic acid 98%
6-Chloro-2-methoxypyridine-3-carboxylic acid
Precaución: for Research Use Only


Formula: C₇H₆ClNO₃
MW: 187.58 g/mol
MDL Number: MFCD11847277
CAS Number: 65515-33-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty