Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

Methyl 3-chloro-2-oxobutanoate 95%

Supplier: AMBEED, INC
Warning

A722901-100MG A722901-250MG
76981-242EA 222 USD InStock
76981-242 76981-244
Methyl 3-chloro-2-oxobutanoate 95%
Methyl 3-chloro-2-oxobutanoate
Precaución: for Research Use Only


Formula: C₅H₇ClO₃
MDL Number: MFCD19707607
CAS Number: 267883-53-4
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty