Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

α,α,α-Trifluoro-p-toluonitrile 98%

Supplier: AMBEED, INC
Danger

A335684-10G A335684-25G
76978-342EA 6 USD InStock
76978-342 76978-344 76978-346
α,α,α-Trifluoro-p-toluonitrile 98%
α,α,α-Trifluoro-p-toluonitrile
Precaución: for Research Use Only


Formula: C₈H₄F₃N
MW: 171.12 g/mol
Flash Pt: 72
MDL Number: MFCD00001826
CAS Number: 455-18-5
UN: 3439
ADR: 6.1 Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty