Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Diphenylmethoxypiperidine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A118588-100MG A118588-1G A118588-250MG A118588-5G
76971-360EA 28 USD InStock
76971-360 76971-362 76971-364 76971-366
4-Diphenylmethoxypiperidine hydrochloride 95%
4-Diphenylmethoxypiperidine hydrochloride


Formula: C₁₈H₂₂ClNO
MDL Number: MFCD16660392
CAS Number: 65214-86-0
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty