Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

4-Methoxymethyl-2,3,5,6-tetrafluorobenzenemethanol 98%

Supplier: AMBEED, INC
Warning

A422839-100G A422839-25G
76979-252EA 88 USD InStock
76979-252 76979-254
4-Methoxymethyl-2,3,5,6-tetrafluorobenzenemethanol 98%
4-Methoxymethyl-2,3,5,6-tetrafluorobenzenemethanol
Precaución: for Research Use Only


Formula: C₉H₈F₄O₂
MDL Number: MFCD08688060
CAS Number: 83282-91-1
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty