Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

6-Nitro-1H-indazol-3-amine 98%

Supplier: AMBEED, INC
Danger

A540559-100MG A540559-250MG
76980-178EA 20 USD InStock
76980-178 76980-180
6-Nitro-1H-indazol-3-amine 98%
6-Nitro-1H-indazol-3-amine
Precaución: for Research Use Only


Formula: C₇H₆N₄O₂
MDL Number: MFCD11707246
CAS Number: 1027259-01-3
UN: 2811
ADR: 6.1,Ⅲ
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty