Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

(S)-3-(Boc-amino)-3-(4-fluorophenyl)propionic acid 98%

Supplier: AMBEED, INC
Warning

A305237-1G A305237-250MG A305237-5G
76977-780EA 16.98 USD InStock
76977-780 76977-782 76977-784 76977-786
(S)-3-(Boc-amino)-3-(4-fluorophenyl)propionic acid 98%
(S)-3-(Boc-amino)-3-(4-fluorophenyl)propionic acid
Precaución: for Research Use Only


Formula: C₁₄H₁₈FNO₄
MDL Number: MFCD03427919
CAS Number: 479064-88-5
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty