Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

trans-N-Boc-1,4-cyclohexanediamine hydrochloride 95%

Supplier: AMBEED, INC
Warning

A768377-1G A768377-250MG A768377-5G
76981-476EA 26 USD InStock
76981-476 76981-478 76981-480
trans-N-Boc-1,4-cyclohexanediamine hydrochloride 95%
trans-N-Boc-1,4-cyclohexanediamine hydrochloride
Precaución: for Research Use Only


Formula: C₁₁H₂₃ClN₂O₂
MDL Number: MFCD04973129
CAS Number: 946002-43-3
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty