Keep my session open?
Ending In 
The session is expired
Your session has expired. For your security, we have logged you out.
Would you like to log in again?

Update to Avantor’s response to the coronavirus (COVID-19) pandemic

3-Fluoro-4-nitrophenylboronic acid pinacol ester 98%

Supplier: AMBEED, INC
Warning

A564752-100MG A564752-1G A564752-250MG A564752-5G
76980-280EA 21 USD InStock
76980-280 76980-282 76980-284 76980-286
3-Fluoro-4-nitrophenylboronic acid pinacol ester 98%
3-Fluoro-4-nitrophenylboronic acid pinacol ester
Precaución: for Research Use Only


Formula: C₁₂H₁₅BFNO₄
MW: 267.06 g/mol
MDL Number: MFCD12026073
CAS Number: 939968-60-2
Order Now

 

ORDER

SPECIFICATIONS

Enter Certificate Details

Enter the Lot number:

Choose from recent batch/lot numbers:

Or
Review & Compare Alternatives
We found alternative products that can save you up to  per item-unit.  To compare product details, select up to 3 alternatives below and click Compare Selected. To add items to your basket, enter a quantity and click Add to Basket.

Original Product:

Description Catalog Number Availability Unit Your Price Price Per Qty